Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTCCCAAAAGGAACGCCTGTGTC[A/T]GCTCACCTTGGGACCGAAATGGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114078 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMK2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAMK2A - calcium/calmodulin dependent protein kinase II alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015981.3 | 2714 | UTR 3 | NP_057065.2 | |||
NM_171825.2 | 2714 | UTR 3 | NP_741960.1 | |||
XM_017009898.1 | 2714 | Intron | XP_016865387.1 |