Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTCATCGCGCTACCGGGTGTGG[A/G]CGACTCCTATCCTCCCCAGAAAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609963 | ||||||||||||||||||||
Literature Links: |
CHSY3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHSY3 - chondroitin sulfate synthase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175856.4 | 1063 | Missense Mutation | GAC,GGC | D,G 230 | NP_787052.3 | |
XM_005271982.3 | 1063 | Missense Mutation | GAC,GGC | D,G 230 | XP_005272039.1 | |
XM_005271983.3 | 1063 | Missense Mutation | GAC,GGC | D,G 230 | XP_005272040.1 | |
XM_011543363.2 | 1063 | Intron | XP_011541665.1 | |||
XM_011543364.2 | 1063 | Intron | XP_011541666.1 | |||
XM_011543365.2 | 1063 | Missense Mutation | GAC,GGC | D,G 230 | XP_011541667.1 | |
XM_017009434.1 | 1063 | Intron | XP_016864923.1 | |||
XM_017009435.1 | 1063 | Missense Mutation | GAC,GGC | D,G 230 | XP_016864924.1 |