Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGAACTTTACAAATTCCACTTTC[C/T]TCCATTAGTGACAGGCCACAGATCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602031 | ||||||||||||||||||||
Literature Links: |
PGGT1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PGGT1B - protein geranylgeranyltransferase type I subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005023.3 | 1032 | Intron | NP_005014.2 | |||
XM_005272020.3 | 1032 | Silent Mutation | GAA,GAG | E,E 333 | XP_005272077.2 | |
XM_011543490.2 | 1032 | Silent Mutation | GAA,GAG | E,E 333 | XP_011541792.1 | |
XM_011543492.2 | 1032 | Silent Mutation | GAA,GAG | E,E 210 | XP_011541794.1 |