Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAAATTTTGCAAGCAATCATAT[G/T]CAGCTCCAGAAGTATGCTCGCCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610607 | ||||||||||||||||||||
Literature Links: |
CPEB4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CPEB4 - cytoplasmic polyadenylation element binding protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308189.1 | 2435 | Missense Mutation | ATG,ATT | M,I 343 | NP_001295118.1 | |
NM_001308191.1 | 2435 | Missense Mutation | ATG,ATT | M,I 343 | NP_001295120.1 | |
NM_001308192.1 | 2435 | Intron | NP_001295121.1 | |||
NM_001308193.1 | 2435 | Intron | NP_001295122.1 | |||
NM_030627.3 | 2435 | Missense Mutation | ATG,ATT | M,I 343 | NP_085130.2 | |
XM_005265994.1 | 2435 | Missense Mutation | ATG,ATT | M,I 343 | XP_005266051.1 | |
XM_011534660.2 | 2435 | Missense Mutation | ATG,ATT | M,I 343 | XP_011532962.1 | |
XM_011534661.2 | 2435 | Missense Mutation | ATG,ATT | M,I 343 | XP_011532963.1 |