Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACCCAAACTAACAAGACTCTGAT[C/T]ATTCAAGGAAAGAAGGCCCAGATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601411 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SGCD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SGCD - sarcoglycan delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000337.5 | Intron | NP_000328.2 | ||||
NM_001128209.1 | Intron | NP_001121681.1 | ||||
NM_172244.2 | Intron | NP_758447.1 | ||||
XM_005265966.4 | Intron | XP_005266023.1 | ||||
XM_005265967.2 | Intron | XP_005266024.1 | ||||
XM_011534621.2 | Intron | XP_011532923.1 | ||||
XM_017009723.1 | Intron | XP_016865212.1 | ||||
XM_017009724.1 | Intron | XP_016865213.1 |