Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGAGAGGAAACTGCTGAGGTTAA[A/T]TTGTTACTGGTGGCTACTGATGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606307 MIM: 606308 MIM: 606309 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCDHA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCDHA1 - protocadherin alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018900.3 | 733 | Intron | NP_061723.1 | |||
NM_031410.2 | 733 | Intron | NP_113598.1 | |||
NM_031411.2 | 733 | Intron | NP_113599.1 |
PCDHA2 - protocadherin alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018905.2 | 733 | Missense Mutation | AAA,AAT | K,N 209 | NP_061728.1 | |
NM_031495.1 | 733 | Missense Mutation | AAA,AAT | K,N 209 | NP_113683.1 | |
NM_031496.1 | 733 | Missense Mutation | AAA,AAT | K,N 209 | NP_113684.1 |
PCDHA3 - protocadherin alpha 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |