Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTGGGGCAGTTTATCCAGGAAG[C/T]GAATGTTGGGGTCAGCCATACTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612374 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM173 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM173 - transmembrane protein 173 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301738.1 | 923 | Missense Mutation | NP_001288667.1 | |||
NM_198282.3 | 923 | Missense Mutation | NP_938023.1 | |||
XM_005268445.3 | 923 | Missense Mutation | XP_005268502.1 | |||
XM_011537639.2 | 923 | Missense Mutation | XP_011535941.1 | |||
XM_011537640.2 | 923 | Missense Mutation | XP_011535942.1 |