Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGAGAAGAGGAGGGCCTGGGGCC[C/T]GTAGAGGCGCTGGAATAGGACGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604660 MIM: 603951 | ||||||||||||||||||||
Literature Links: |
KCNIP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNIP1 - potassium voltage-gated channel interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001034837.2 | 900 | Intron | NP_001030009.1 | |||
NM_001034838.2 | 900 | Intron | NP_001030010.1 | |||
NM_001278339.1 | 900 | Intron | NP_001265268.1 | |||
NM_001278340.1 | 900 | Intron | NP_001265269.1 | |||
NM_014592.3 | 900 | Intron | NP_055407.1 | |||
XM_017009407.1 | 900 | Intron | XP_016864896.1 | |||
XM_017009408.1 | 900 | Intron | XP_016864897.1 |
KCNMB1 - potassium calcium-activated channel subfamily M regulatory beta subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004137.3 | 900 | Missense Mutation | NP_004128.1 |