Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTTCTTTAAACAAATAATATAG[C/T]TTATTTTTCTGATATGCTCTATTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171833 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIK3R1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIK3R1 - phosphoinositide-3-kinase regulatory subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242466.1 | Intron | NP_001229395.1 | ||||
NM_181504.3 | Intron | NP_852556.2 | ||||
NM_181523.2 | Intron | NP_852664.1 | ||||
NM_181524.1 | Intron | NP_852665.1 | ||||
XM_005248542.3 | Intron | XP_005248599.1 | ||||
XM_011543493.2 | Intron | XP_011541795.1 | ||||
XM_017009585.1 | Intron | XP_016865074.1 | ||||
XM_017009586.1 | Intron | XP_016865075.1 |