Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTAATAATCTGATAATCGGCCAG[A/G]TCAGGGTAGACTCTGAAAACTCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
STK32A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STK32A - serine/threonine kinase 32A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001112724.1 | Intron | NP_001106195.1 | ||||
NM_001287740.1 | Intron | NP_001274669.1 | ||||
NM_145001.3 | Intron | NP_659438.1 | ||||
XM_011537576.1 | Intron | XP_011535878.1 | ||||
XM_011537577.2 | Intron | XP_011535879.1 | ||||
XM_011537579.2 | Intron | XP_011535881.1 | ||||
XM_017009213.1 | Intron | XP_016864702.1 | ||||
XM_017009214.1 | Intron | XP_016864703.1 |