Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTCTGAGCTGAAATACAGACTG[C/T]GGTGTTAATATCCTTTTCTTCTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605675 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF14 - ring finger protein 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201365.1 | 566 | UTR 5 | NP_001188294.1 | |||
NM_004290.4 | 566 | UTR 5 | NP_004281.1 | |||
NM_183398.2 | 566 | UTR 5 | NP_899645.1 | |||
NM_183399.2 | 566 | Intron | NP_899646.1 | |||
NM_183400.2 | 566 | Intron | NP_899647.1 | |||
NM_183401.2 | 566 | UTR 5 | NP_899648.1 | |||
XM_005268536.2 | 566 | UTR 5 | XP_005268593.1 | |||
XM_005268537.2 | 566 | UTR 5 | XP_005268594.1 | |||
XM_005268539.2 | 566 | UTR 5 | XP_005268596.1 | |||
XM_005268540.3 | 566 | UTR 5 | XP_005268597.1 | |||
XM_005268541.3 | 566 | Intron | XP_005268598.1 | |||
XM_005268542.2 | 566 | UTR 5 | XP_005268599.1 | |||
XM_005268543.4 | 566 | Intron | XP_005268600.1 | |||
XM_011537714.2 | 566 | Intron | XP_011536016.1 | |||
XM_017010070.1 | 566 | UTR 5 | XP_016865559.1 | |||
XM_017010071.1 | 566 | UTR 5 | XP_016865560.1 | |||
XM_017010072.1 | 566 | UTR 5 | XP_016865561.1 | |||
XM_017010073.1 | 566 | UTR 5 | XP_016865562.1 | |||
XM_017010074.1 | 566 | UTR 5 | XP_016865563.1 | |||
XM_017010075.1 | 566 | UTR 5 | XP_016865564.1 | |||
XM_017010076.1 | 566 | UTR 5 | XP_016865565.1 | |||
XM_017010077.1 | 566 | Intron | XP_016865566.1 |