Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACAATGTTCCGACCACATGAAAA[A/G]AAGTTACCTATTAAAGAGACTCTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM267 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM267 - transmembrane protein 267 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022483.4 | Intron | NP_071928.2 | ||||
XM_005248333.4 | Intron | XP_005248390.1 | ||||
XM_005248335.3 | Intron | XP_005248392.1 | ||||
XM_006714486.3 | Intron | XP_006714549.1 | ||||
XM_011514074.2 | Intron | XP_011512376.1 | ||||
XM_011514075.2 | Intron | XP_011512377.1 |