Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTTATAGATCACTTTTTAATTC[A/G]GGCCTCTGCAGCATTAGAAAAATTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C5orf51 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C5orf51 - chromosome 5 open reading frame 51 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175921.5 | 269 | Missense Mutation | CAG,CGG | Q,R 39 | NP_787117.3 | |
XM_005248289.4 | 269 | Missense Mutation | CAG,CGG | Q,R 39 | XP_005248346.1 | |
XM_011514032.2 | 269 | UTR 5 | XP_011512334.1 |