Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGTAGGCGCAAGCTGGAAAGTGC[A/C]AGTCCTGTGGCTTCCAGAGGCAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 158120 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD14 - CD14 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000591.3 | 818 | Silent Mutation | CTG,CTT | L,L 142 | NP_000582.1 | |
NM_001040021.2 | 818 | Silent Mutation | CTG,CTT | L,L 142 | NP_001035110.1 | |
NM_001174104.1 | 818 | Silent Mutation | CTG,CTT | L,L 142 | NP_001167575.1 | |
NM_001174105.1 | 818 | Silent Mutation | CTG,CTT | L,L 142 | NP_001167576.1 |
TMCO6 - transmembrane and coiled-coil domains 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300980.1 | 818 | Intron | NP_001287909.1 | |||
NM_001300982.1 | 818 | Intron | NP_001287911.1 | |||
NM_018502.4 | 818 | Intron | NP_060972.3 | |||
XM_005268477.1 | 818 | Intron | XP_005268534.1 | |||
XM_011537663.2 | 818 | Intron | XP_011535965.1 | |||
XM_011537665.2 | 818 | Intron | XP_011535967.1 | |||
XM_011537668.2 | 818 | Intron | XP_011535970.1 | |||
XM_017009617.1 | 818 | Intron | XP_016865106.1 | |||
XM_017009618.1 | 818 | Intron | XP_016865107.1 | |||
XM_017009619.1 | 818 | Intron | XP_016865108.1 | |||
XM_017009620.1 | 818 | Intron | XP_016865109.1 |