Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTCTGCACTCAAAAGAATTTTGC[A/G]TGGAAGGAAGAATATGATACACGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJC21 - DnaJ heat shock protein family (Hsp40) member C21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012339.2 | 722 | Silent Mutation | GCA,GCG | A,A 165 | NP_001012339.2 | |
NM_194283.3 | 722 | Silent Mutation | GCA,GCG | A,A 165 | NP_919259.3 | |
XM_005248249.4 | 722 | Silent Mutation | GCA,GCG | A,A 252 | XP_005248306.2 | |
XM_005248250.3 | 722 | Silent Mutation | GCA,GCG | A,A 252 | XP_005248307.2 | |
XM_011513965.2 | 722 | Silent Mutation | GCA,GCG | A,A 252 | XP_011512267.2 | |
XM_011513966.2 | 722 | Silent Mutation | GCA,GCG | A,A 252 | XP_011512268.2 |