Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCCTCAAGGGCTGAGGGCAGAG[A/T]TGGGGTCTGGGGGGCAGCCAGGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126660 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DBN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DBN1 - drebrin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004395.3 | 1969 | Missense Mutation | ACT,TCT | T,S 561 | NP_004386.2 | |
NM_080881.2 | 1969 | Missense Mutation | ACT,TCT | T,S 563 | NP_543157.1 | |
XM_005265827.3 | 1969 | Missense Mutation | ACT,TCT | T,S 607 | XP_005265884.1 | |
XM_006714826.2 | 1969 | Missense Mutation | ACT,TCT | T,S 544 | XP_006714889.1 | |
XM_011534446.2 | 1969 | Missense Mutation | ACT,TCT | T,S 609 | XP_011532748.1 | |
XM_011534447.2 | 1969 | Missense Mutation | ACT,TCT | T,S 596 | XP_011532749.1 | |
XM_017009139.1 | 1969 | Missense Mutation | ACT,TCT | T,S 609 | XP_016864628.1 | |
XM_017009140.1 | 1969 | Missense Mutation | ACT,TCT | T,S 563 | XP_016864629.1 |
PRR7 - proline rich 7 (synaptic) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |