Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTGGTCCGAAAGCAATTCCTCTC[C/T]GTGTTCAGAGATTCCAGTGCTGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
N4BP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
N4BP3 - NEDD4 binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015111.1 | Intron | NP_055926.1 | ||||
XM_006714834.3 | Intron | XP_006714897.1 | ||||
XM_011534473.1 | Intron | XP_011532775.1 | ||||
XM_011534474.1 | Intron | XP_011532776.1 |