Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606336 MIM: 606345 MIM: 606333 MIM: 606334 MIM: 606335 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCDHB10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCDHB10 - protocadherin beta 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCDHB16 - protocadherin beta 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020957.3 | 1028 | Missense Mutation | GCG,GGG | A,G 53 | NP_066008.2 |
PCDHB7 - protocadherin beta 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCDHB8 - protocadherin beta 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCDHB9 - protocadherin beta 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |