Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATATATTTACTTAGTTTCAAAGA[C/G]CAGGAATCCTGCAAAGGTGGAGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612045 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QTNF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1QTNF3 - C1q and tumor necrosis factor related protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030945.3 | 1084 | Missense Mutation | CTC,GTC | L,V 242 | NP_112207.1 | |
NM_181435.5 | 1084 | Missense Mutation | CTC,GTC | L,V 315 | NP_852100.3 |
C1QTNF3-AMACR - C1QTNF3-AMACR readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |