Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAATTACTATGAATTTGATGTGC[A/G]GGCTCGCGATGGGGGTTCTCCAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606288 MIM: 606297 MIM: 606298 MIM: 603059 MIM: 606289 MIM: 606290 MIM: 606291 MIM: 606292 MIM: 606293 MIM: 606294 MIM: 606295 MIM: 606296 MIM: 606299 MIM: 606300 MIM: 606301 MIM: 603058 MIM: 606302 MIM: 606303 MIM: 606304 MIM: 603627 MIM: 606305 MIM: 606306 | ||||||||||||||||||||
Literature Links: |
PCDHGA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCDHGA1 - protocadherin gamma subfamily A, 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018912.2 | 962 | Intron | NP_061735.1 | |||
NM_031993.1 | 962 | Intron | NP_114382.1 |
PCDHGA10 - protocadherin gamma subfamily A, 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018913.2 | 962 | Intron | NP_061736.1 | |||
NM_032090.1 | 962 | Intron | NP_114479.1 |
PCDHGA11 - protocadherin gamma subfamily A, 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018914.2 | 962 | Intron | NP_061737.1 | |||
NM_032091.1 | 962 | Intron | NP_114480.1 | |||
NM_032092.1 | 962 | Intron | NP_114481.1 |
PCDHGA12 - protocadherin gamma subfamily A, 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003735.2 | 962 | Intron | NP_003726.1 | |||
NM_032094.1 | 962 | Intron | NP_115265.1 |
PCDHGA2 - protocadherin gamma subfamily A, 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018915.3 | 962 | Intron | NP_061738.1 | |||
NM_032009.2 | 962 | Intron | NP_114398.1 |
PCDHGA3 - protocadherin gamma subfamily A, 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018916.3 | 962 | Intron | NP_061739.2 | |||
NM_032011.1 | 962 | Intron | NP_114400.1 |
PCDHGA4 - protocadherin gamma subfamily A, 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018917.3 | 962 | Intron | NP_061740.2 | |||
NM_032053.2 | 962 | Intron | NP_114442.2 |
PCDHGA5 - protocadherin gamma subfamily A, 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018918.2 | 962 | Intron | NP_061741.1 | |||
NM_032054.1 | 962 | Intron | NP_114443.1 |
PCDHGA6 - protocadherin gamma subfamily A, 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018919.2 | 962 | Intron | NP_061742.1 | |||
NM_032086.1 | 962 | Intron | NP_114475.1 |
PCDHGA7 - protocadherin gamma subfamily A, 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018920.3 | 962 | Intron | NP_061743.1 | |||
NM_032087.2 | 962 | Intron | NP_114476.1 |
PCDHGA8 - protocadherin gamma subfamily A, 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014004.2 | 962 | Intron | NP_054723.1 | |||
NM_032088.1 | 962 | Intron | NP_114477.1 |
PCDHGA9 - protocadherin gamma subfamily A, 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018921.2 | 962 | Intron | NP_061744.1 | |||
NM_032089.1 | 962 | Intron | NP_114478.1 |
PCDHGB1 - protocadherin gamma subfamily B, 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018922.2 | 962 | Intron | NP_061745.1 | |||
NM_032095.1 | 962 | Intron | NP_115266.1 |
PCDHGB2 - protocadherin gamma subfamily B, 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018923.2 | 962 | Intron | NP_061746.1 | |||
NM_032096.1 | 962 | Intron | NP_115267.1 |
PCDHGB3 - protocadherin gamma subfamily B, 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018924.4 | 962 | Intron | NP_061747.2 | |||
NM_032097.2 | 962 | Intron | NP_115268.2 |
PCDHGB4 - protocadherin gamma subfamily B, 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003736.2 | 962 | Intron | NP_003727.1 | |||
NM_032098.1 | 962 | Intron | NP_115269.1 |
PCDHGB5 - protocadherin gamma subfamily B, 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018925.2 | 962 | Intron | NP_061748.1 | |||
NM_032099.1 | 962 | Intron | NP_115270.1 |
PCDHGB6 - protocadherin gamma subfamily B, 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018926.2 | 962 | Intron | NP_061749.1 | |||
NM_032100.1 | 962 | Intron | NP_115271.1 |
PCDHGB7 - protocadherin gamma subfamily B, 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018927.3 | 962 | Intron | NP_061750.1 | |||
NM_032101.2 | 962 | Intron | NP_115272.1 |
PCDHGC3 - protocadherin gamma subfamily C, 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002588.3 | 962 | Intron | NP_002579.2 | |||
NM_032402.1 | 962 | Intron | NP_115778.1 | |||
NM_032403.2 | 962 | Intron | NP_115779.1 |
PCDHGC4 - protocadherin gamma subfamily C, 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018928.2 | 962 | Missense Mutation | CAG,CGG | Q,R 321 | NP_061751.1 | |
NM_032406.1 | 962 | Missense Mutation | CAG,CGG | Q,R 321 | NP_115782.1 |
PCDHGC5 - protocadherin gamma subfamily C, 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |