Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCAACAGTTCTCTGTGAGTTTTG[C/G]AGACTGGCATGCTTTTCTGAAAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607005 | ||||||||||||||||||||
Literature Links: |
GEMIN5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GEMIN5 - gem nuclear organelle associated protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252156.1 | 4181 | Silent Mutation | CTC,CTG | L,L 1366 | NP_001239085.1 | |
NM_015465.4 | 4181 | Silent Mutation | CTC,CTG | L,L 1367 | NP_056280.2 |