Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTTAAAATGTATTTAGAGTTTG[C/G]TAAGACTGGTCTGTGTTATTTTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153455 MIM: 610479 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOX - lysyl oxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178102.2 | 3127 | UTR 3 | NP_001171573.1 | |||
NM_001317073.1 | 3127 | UTR 3 | NP_001304002.1 | |||
NM_002317.6 | 3127 | UTR 3 | NP_002308.2 |
SRFBP1 - serum response factor binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152546.2 | 3127 | Intron | NP_689759.2 | |||
XM_017009111.1 | 3127 | Intron | XP_016864600.1 |