Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCTTGTAAATCTTCACTTGCTGG[C/T]ATATCTCGGTTCTCTGGGTGACCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601484 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC152 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC152 - coiled-coil domain containing 152 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134848.1 | 2131 | UTR 3 | NP_001128320.1 |
SEPP1 - selenoprotein P, plasma, 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085486.1 | 2131 | Missense Mutation | ATA,ATG | I,M 263 | NP_001078955.1 | |
NM_001093726.1 | 2131 | Missense Mutation | ATA,ATG | I,M 293 | NP_001087195.1 | |
NM_005410.2 | 2131 | Missense Mutation | ATA,ATG | I,M 263 | NP_005401.3 |