Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACTGGCCTCTAGAGATTAACCTC[C/T]TGTATTCAATTTCCAAACCAAATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615931 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRR16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRR16 - proline rich 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300783.1 | Intron | NP_001287712.1 | ||||
NM_001308087.1 | Intron | NP_001295016.1 | ||||
NM_001308088.1 | Intron | NP_001295017.1 | ||||
NM_016644.2 | Intron | NP_057728.1 | ||||
XM_011543452.2 | Intron | XP_011541754.1 | ||||
XM_011543453.2 | Intron | XP_011541755.1 | ||||
XM_011543454.2 | Intron | XP_011541756.1 |