Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGGTTTGTGGAATTGGGTGTGTT[C/T]GGCACTGGAAACACCGTGTTGCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GAPT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GAPT - GRB2-binding adaptor protein, transmembrane | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304428.1 | 616 | Missense Mutation | CGG,TGG | R,W 31 | NP_001291357.1 | |
NM_001304429.1 | 616 | Missense Mutation | CGG,TGG | R,W 31 | NP_001291358.1 | |
NM_001304431.1 | 616 | Missense Mutation | CGG,TGG | R,W 31 | NP_001291360.1 | |
NM_152687.3 | 616 | Missense Mutation | CGG,TGG | R,W 31 | NP_689900.1 |