Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGGTGCTTATGATGTTAAAACT[G/T]TAGAAGTATTGAAAGGACCAGTATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600936 | ||||||||||||||||||||
Literature Links: |
HMMR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HMMR - hyaluronan mediated motility receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142556.1 | 267 | Missense Mutation | GTA,TTA | V,L 29 | NP_001136028.1 | |
NM_001142557.1 | 267 | Intron | NP_001136029.1 | |||
NM_012484.2 | 267 | Missense Mutation | GTA,TTA | V,L 29 | NP_036616.2 | |
NM_012485.2 | 267 | Missense Mutation | GTA,TTA | V,L 29 | NP_036617.2 |
NUDCD2 - NudC domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |