Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGTGGTGTTACTGGATGCAGCA[A/G]GACTTGGATGTAGTTGCACCCCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 180071 MIM: 608886 | ||||||||||||||||||||
Literature Links: |
PDE6A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDE6A - phosphodiesterase 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000440.2 | 1757 | Silent Mutation | TCC,TCT | S,S 856 | NP_000431.2 | |
XM_011537650.2 | 1757 | Silent Mutation | TCC,TCT | S,S 561 | XP_011535952.1 | |
XM_011537651.2 | 1757 | Silent Mutation | TCC,TCT | S,S 507 | XP_011535953.1 | |
XM_011537652.1 | 1757 | Silent Mutation | TCC,TCT | S,S 497 | XP_011535954.1 | |
XM_011537653.2 | 1757 | Silent Mutation | TCC,TCT | S,S 497 | XP_011535955.1 | |
XM_011537654.2 | 1757 | Silent Mutation | TCC,TCT | S,S 497 | XP_011535956.1 | |
XM_017009572.1 | 1757 | Silent Mutation | TCC,TCT | S,S 775 | XP_016865061.1 |
PPARGC1B - PPARG coactivator 1 beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |