Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGCTGAACCGGTGTGGCGAGGC[A/G]GCGCGGAGCCTGCCCCTGGGCGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611842 MIM: 603848 | ||||||||||||||||||||
Literature Links: |
MRPL36 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRPL36 - mitochondrial ribosomal protein L36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032479.3 | 87 | Intron | NP_115868.1 | |||
XM_011514079.2 | 87 | Intron | XP_011512381.1 | |||
XM_011514080.2 | 87 | Intron | XP_011512382.1 | |||
XM_017009750.1 | 87 | Intron | XP_016865239.1 | |||
XM_017009751.1 | 87 | Intron | XP_016865240.1 | |||
XM_017009752.1 | 87 | Intron | XP_016865241.1 |
NDUFS6 - NADH:ubiquinone oxidoreductase subunit S6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004553.4 | 87 | Silent Mutation | GCA,GCG | A,A 17 | NP_004544.1 |