Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGGCTTCTAACTTCACTGAATC[C/T]AGTACTAAACGTGCTTCTTTATATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610058 | ||||||||||||||||||||
Literature Links: |
TBCA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TBCA - tubulin folding cofactor A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297738.1 | 407 | UTR 3 | NP_001284667.1 | |||
NM_001297740.1 | 407 | Silent Mutation | CTA,CTG | L,L 72 | NP_001284669.1 | |
NM_004607.2 | 407 | Silent Mutation | CTA,CTG | L,L 101 | NP_004598.1 |