Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGATCACCACGACGACAACGGTCC[C/T]GGCTGTGTTGACCAGAAAGCCCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609445 MIM: 606202 | ||||||||||||||||||||
Literature Links: |
RXFP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RXFP3 - relaxin/insulin like family peptide receptor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC45A2 - solute carrier family 45 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012509.3 | 1596 | Intron | NP_001012527.1 | |||
NM_001297417.2 | 1596 | Intron | NP_001284346.2 | |||
NM_016180.4 | 1596 | Missense Mutation | AGG,GGG | R,G 502 | NP_057264.3 |