Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCAGTTCACCTTCACCGCCGACC[G/T]GCCGCAGCTGCACTGCGCAGCCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 122559 MIM: 610103 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRHBP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRHBP - corticotropin releasing hormone binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001882.3 | 507 | Missense Mutation | CGG,CTG | R,L 76 | NP_001873.2 |
S100Z - S100 calcium binding protein Z | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |