Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAATGTACATAGGGTCCAGCTGAC[A/G]CTTTTCCCACTCCCCCATCAGTAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606698 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf52 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C5orf52 - chromosome 5 open reading frame 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145132.1 | Intron | NP_001138604.1 | ||||
XM_011534416.1 | Intron | XP_011532718.1 | ||||
XM_017008909.1 | Intron | XP_016864398.1 |
SOX30 - SRY-box 30 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |