Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGTTGGCCTCTGGGTTACTGAT[A/G]TCCCATAAGTGAGAAGAAGGTCATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610890 | ||||||||||||||||||||
Literature Links: |
RGS7BP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RGS7BP - regulator of G-protein signaling 7 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029875.2 | 839 | Intron | NP_001025046.1 | |||
NM_001271890.1 | 839 | Intron | NP_001258819.1 | |||
NM_001271891.1 | 839 | Intron | NP_001258820.1 | |||
XM_005248502.3 | 839 | UTR 5 | XP_005248559.1 |