Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCCTGCTGGGTAGGGAAGGCTC[C/T]GTAGGGAAGGCAGGGAGGCTTGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF454 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF454 - zinc finger protein 454 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178089.2 | Intron | NP_001171560.1 | ||||
NM_001178090.2 | Intron | NP_001171561.1 | ||||
NM_001323306.1 | Intron | NP_001310235.1 | ||||
NM_001323307.1 | Intron | NP_001310236.1 | ||||
NM_001323308.1 | Intron | NP_001310237.1 | ||||
NM_001323309.1 | Intron | NP_001310238.1 | ||||
NM_001323310.1 | Intron | NP_001310239.1 | ||||
NM_182594.3 | Intron | NP_872400.2 |