Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGTGACGACTCCCCCAGATATC[A/C]CCGGCAGCCGCAACCTGCACTGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610826 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A46 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC25A46 - solute carrier family 25 member 46 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM232 - transmembrane protein 232 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039763.3 | Intron | NP_001034852.3 | ||||
XM_006714670.3 | Intron | XP_006714733.1 | ||||
XM_011543552.2 | Intron | XP_011541854.1 | ||||
XM_011543553.2 | Intron | XP_011541855.1 | ||||
XM_011543555.2 | Intron | XP_011541857.1 | ||||
XM_011543556.1 | Intron | XP_011541858.1 | ||||
XM_011543557.2 | Intron | XP_011541859.1 | ||||
XM_011543559.2 | Intron | XP_011541861.1 | ||||
XM_011543560.2 | Intron | XP_011541862.1 | ||||
XM_011543561.2 | Intron | XP_011541863.1 | ||||
XM_011543563.2 | Intron | XP_011541865.1 | ||||
XM_011543564.2 | Intron | XP_011541866.1 | ||||
XM_011543565.2 | Intron | XP_011541867.1 | ||||
XM_011543566.2 | Intron | XP_011541868.1 | ||||
XM_011543567.2 | Intron | XP_011541869.1 | ||||
XM_017009704.1 | Intron | XP_016865193.1 | ||||
XM_017009705.1 | Intron | XP_016865194.1 | ||||
XM_017009706.1 | Intron | XP_016865195.1 | ||||
XM_017009707.1 | Intron | XP_016865196.1 |