Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGAGAGGATCTTTTTCTTTCCT[A/G]TAAGTGCTTTGCCGTCTTCTGAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD34B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD34B - ankyrin repeat domain 34B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004441.2 | 1602 | Missense Mutation | ACA,ATA | T,I 391 | NP_001004441.2 | |
XM_006714599.3 | 1602 | Missense Mutation | ACA,ATA | T,I 391 | XP_006714662.1 | |
XM_011543366.2 | 1602 | Missense Mutation | ACA,ATA | T,I 391 | XP_011541668.1 | |
XM_011543369.2 | 1602 | Missense Mutation | ACA,ATA | T,I 391 | XP_011541671.1 |