Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAAAGCATATTTAGTGCTGTAGT[C/G]TGAGATTCTGATGGATTTTCATCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114090 MIM: 606832 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAST PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERAP1 - endoplasmic reticulum aminopeptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040458.1 | 2948 | Intron | NP_001035548.1 | |||
NM_001198541.1 | 2948 | Intron | NP_001185470.1 | |||
NM_016442.3 | 2948 | UTR 3 | NP_057526.3 | |||
XM_005272015.4 | 2948 | Intron | XP_005272072.1 | |||
XM_005272016.4 | 2948 | Intron | XP_005272073.1 | |||
XM_011543480.2 | 2948 | Intron | XP_011541782.1 | |||
XM_011543481.2 | 2948 | Intron | XP_011541783.1 | |||
XM_011543484.2 | 2948 | Intron | XP_011541786.1 | |||
XM_011543485.2 | 2948 | Intron | XP_011541787.1 | |||
XM_011543486.2 | 2948 | Intron | XP_011541788.1 | |||
XM_017009580.1 | 2948 | Intron | XP_016865069.1 | |||
XM_017009581.1 | 2948 | Intron | XP_016865070.1 | |||
XM_017009582.1 | 2948 | UTR 3 | XP_016865071.1 | |||
XM_017009583.1 | 2948 | Intron | XP_016865072.1 |