Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTTGCAAATCTCGATTTACTGAG[A/G]ATGGGTGTACTTGTGCTTTGCCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612720 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHX29 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHX29 - DEAH-box helicase 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019030.2 | 3733 | Missense Mutation | TCC,TTC | S,F 1259 | NP_061903.2 | |
XM_006714653.2 | 3733 | Missense Mutation | TCC,TTC | S,F 1208 | XP_006714716.1 | |
XM_017009588.1 | 3733 | Missense Mutation | TCC,TTC | S,F 623 | XP_016865077.1 |