Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGGCTTCTTTAGTGCCCACATT[A/G]GCAGCTGTGGTTTCGGTTTGGGTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616073 | ||||||||||||||||||||
Literature Links: |
DEPDC1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DEPDC1B - DEP domain containing 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145208.1 | 1595 | Missense Mutation | CCA,CTA | P,L 448 | NP_001138680.1 | |
NM_018369.2 | 1595 | Missense Mutation | CCA,CTA | P,L 510 | NP_060839.2 | |
XM_011543509.2 | 1595 | Missense Mutation | CCA,CTA | P,L 495 | XP_011541811.1 | |
XM_011543510.2 | 1595 | Intron | XP_011541812.1 | |||
XM_017009629.1 | 1595 | Intron | XP_016865118.1 |