Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAAGACGTATTTCCGCCGCTTG[C/T]TGCGGGTGGGCTGGCAGCACTGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603745 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLIT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLIT3 - slit guidance ligand 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271946.1 | 4895 | Missense Mutation | AAC,AGC | N,S 1499 | NP_001258875.1 | |
NM_003062.3 | 4895 | Missense Mutation | AAC,AGC | N,S 1492 | NP_003053.1 | |
XM_017009779.1 | 4895 | Missense Mutation | AAC,AGC | N,S 1429 | XP_016865268.1 |