Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGTGGCTTTCTTCATTTTATTGA[G/T]GCTTTGGGGAGAGGTGGGTTCTGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606332 | ||||||||||||||||||||
Literature Links: |
PCDHB17P PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCDHB17P - protocadherin beta 17 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCDHB6 - protocadherin beta 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303145.1 | 295 | Intron | NP_001290074.1 | |||
NM_018939.3 | 295 | Missense Mutation | AGG,ATG | R,M 20 | NP_061762.2 |