Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCACTCTTCTTATGCTGTCTGCG[C/T]AACTGTGAGCCAATTAAACCTCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf64 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C5orf64 - chromosome 5 open reading frame 64 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173667.3 | 136 | Silent Mutation | CGC,CGT | R,R 11 | NP_775938.1 | |
XM_017009378.1 | 136 | Intron | XP_016864867.1 | |||
XM_017009379.1 | 136 | Intron | XP_016864868.1 | |||
XM_017009380.1 | 136 | Intron | XP_016864869.1 | |||
XM_017009381.1 | 136 | Intron | XP_016864870.1 | |||
XM_017009382.1 | 136 | Intron | XP_016864871.1 |