Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGTACTGGTCACCGGCGCGGGGG[C/G]AGGTGAGCATGCGAAGGTTGGAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601860 | ||||||||||||||||||||
Literature Links: |
HSD17B4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HSD17B4 - hydroxysteroid 17-beta dehydrogenase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000414.3 | 189 | Missense Mutation | GCA,GGA | A,G 19 | NP_000405.1 | |
NM_001199291.2 | 189 | UTR 5 | NP_001186220.1 | |||
NM_001199292.1 | 189 | Missense Mutation | GCA,GGA | A,G 19 | NP_001186221.1 | |
NM_001292027.1 | 189 | UTR 5 | NP_001278956.1 | |||
NM_001292028.1 | 189 | UTR 5 | NP_001278957.1 |