Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGATAAGACTCTGCCACTCCAGA[C/G]GAGTTCCTTATGTCAGAGGGGAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601535 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFNA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFNA5 - ephrin A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001962.2 | 4480 | UTR 3 | NP_001953.1 | |||
XM_006714565.2 | 4480 | UTR 3 | XP_006714628.1 | |||
XM_011543250.2 | 4480 | UTR 3 | XP_011541552.1 | |||
XM_011543251.2 | 4480 | UTR 3 | XP_011541553.1 | |||
XM_017009205.1 | 4480 | UTR 3 | XP_016864694.1 |