Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATGGCTTCACCGGCCTGTCCTCC[A/G]TGTTCCTGATAGCGACCCCGGAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603377 | ||||||||||||||||||||
Literature Links: |
LOC553103 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC553103 - uncharacterized LOC553103 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3936 - microRNA 3936 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC22A5 - solute carrier family 22 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308122.1 | 382 | Missense Mutation | ATG,GTG | M,V 40 | NP_001295051.1 | |
NM_003060.3 | 382 | Missense Mutation | ATG,GTG | M,V 40 | NP_003051.1 | |
XM_011543590.2 | 382 | Intron | XP_011541892.1 | |||
XM_017009778.1 | 382 | Intron | XP_016865267.1 |