Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATACTTTTGACTCATACAAATATA[C/T]GTTTAAAACTATTACCTACAAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 617016 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLCXD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLCXD3 - phosphatidylinositol specific phospholipase C X domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005473.2 | 5518 | UTR 3 | NP_001005473.1 | |||
XM_017009436.1 | 5518 | Intron | XP_016864925.1 | |||
XM_017009437.1 | 5518 | Intron | XP_016864926.1 | |||
XM_017009438.1 | 5518 | Intron | XP_016864927.1 |