Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGGCAGGGAAGTCCAGGCCCTT[A/G]GAGGCAACGTCTGTGGCTACTAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608170 MIM: 611435 MIM: 615813 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX41 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX41 - DEAD-box helicase 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321732.1 | 1860 | Silent Mutation | TCC,TCT | S,S 367 | NP_001308661.1 | |
NM_001321830.1 | 1860 | Silent Mutation | TCC,TCT | S,S 367 | NP_001308759.1 | |
NM_016222.3 | 1860 | Silent Mutation | TCC,TCT | S,S 493 | NP_057306.2 |
DOK3 - docking protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM193B - family with sequence similarity 193 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |