Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTGTGTCCAAAGGGAACAAACAG[C/T]ATTTTCAGCTCAGTCATCAGACAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606328 MIM: 606329 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCDHB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCDHB2 - protocadherin beta 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCDHB3 - protocadherin beta 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018937.4 | 628 | Missense Mutation | CAT,TAT | H,Y 74 | NP_061760.1 |