Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTACCTGTGGAGGCAACACTGG[C/T]CAAGAAGAGGAAAGGTAGCTGCAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611502 | ||||||||||||||||||||
Literature Links: |
CENPK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPWD1 - peptidylprolyl isomerase domain and WD repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278926.1 | 254 | Missense Mutation | CCA,TCA | P,S 19 | NP_001265855.1 | |
NM_001278927.1 | 254 | UTR 5 | NP_001265856.1 | |||
NM_001278929.1 | 254 | UTR 5 | NP_001265858.1 | |||
NM_015342.3 | 254 | Missense Mutation | GCC,GTC | A,V 61 | NP_056157.1 | |
XM_005248474.3 | 254 | Missense Mutation | CCA,TCA | P,S 19 | XP_005248531.1 | |
XM_006714583.3 | 254 | UTR 5 | XP_006714646.1 | |||
XM_011543295.1 | 254 | Intron | XP_011541597.1 | |||
XM_017009286.1 | 254 | UTR 5 | XP_016864775.1 |