Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTGGGTGGTGGCTCAGGAATATC[C/G]AGAGTCTCTATGTCGCTCTTTTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610174 | ||||||||||||||||||||
Literature Links: |
EFCAB9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EFCAB9 - EF-hand calcium binding domain 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBTD2 - ubiquitin domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152277.2 | 543 | Silent Mutation | CTC,CTG | L,L 139 | NP_689490.2 | |
XM_017010022.1 | 543 | Silent Mutation | CTC,CTG | L,L 95 | XP_016865511.1 | |
XM_017010023.1 | 543 | Silent Mutation | CTC,CTG | L,L 95 | XP_016865512.1 |